Gene Transcripts

Gene Symbol
Rwdd2b
Entrez Gene ID
53858
Gene Name
RWD domain containing 2B
Gene Aliases
ORF5
Chromosome Location
Chr. 16: 87433331 - 87440592 on Build GRCm38
UniGene ID
Mm.162406
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016924.2 NP_058620.2 5 840
GenBank mRNA AF177771.1 - 5 796
AK002746.1 - 5 806
AK084966.1 - 5 815
BC002143.1 - 5 821 3484719

View Full Product Details