Gene Transcripts

Gene Symbol
Hebp2
Entrez Gene ID
56016
Gene Name
heme binding protein 2
Gene Aliases
SOUL
Chromosome Location
Chr. 10: 18540123 - 18546076 on Build GRCm38
UniGene ID
Mm.35551
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019487.3 NP_062360.1 4 859
GenBank mRNA AF117614.1 - 4 789
AK078041.1 - 843
AK078130.1 - 4 859
BC006898.1 - 4 552 3599858

View Full Product Details