Gene Transcripts
- Gene Symbol
- Pdcd10
- Entrez Gene ID
- 56426
- Gene Name
- programmed cell death 10
- Gene Aliases
- 2410003B13Rik, Ccm3, Tfa15, Tfar15
- Chromosome Location
- Chr. 3: 75516490 - 75556852 on Build GRCm38
- UniGene ID
- Mm.316473
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019745.4 |
NP_062719.2
|
4, 5 |
523 |
|
GenBank mRNA |
AF159368.1 |
- |
4, 5 |
426 |
|
AK005594.1 |
- |
4, 5 |
452 |
|
AK075926.1 |
- |
4, 5 |
471 |
|
AK077308.1 |
- |
4, 5 |
500 |
|
AK137198.1 |
- |
4, 5 |
493 |
|
AK149182.1 |
- |
4, 5 |
429 |
|
AK160234.1 |
- |
4, 5 |
473 |
|
AK168857.1 |
- |
4, 5 |
457 |
|
AK169689.1 |
- |
4, 5 |
457 |
|
BC019650.1 |
- |
4, 5 |
463 |
4022513 |
BC086778.1 |
- |
4, 5 |
442 |
5689088 |
View Full Product Details