Gene Transcripts
- Gene Symbol
- Chn1
- Entrez Gene ID
- 84030
- Gene Name
- chimerin 1
- Gene Aliases
- Chromosome Location
- Chr. 3: 60512360 - 60668281 on Build Rnor_6.0
- UniGene ID
- Rn.11166
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_032083.1 |
NP_114472.1
|
7 |
2227 |
|
GenBank mRNA |
X67250.1 |
- |
7 |
2227 |
|
View Full Product Details