Gene Transcripts
- Gene Symbol
- PDCD10
- Entrez Gene ID
- 11235
- Gene Name
- programmed cell death 10
- Gene Aliases
- CCM3, TFAR15
- Chromosome Location
- Chr. 3: 167683891 - 167735690 on Build GRCh38
- UniGene ID
- Hs.478150
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_007217.3 |
NP_009148.2
|
4, 5 |
556 |
|
NM_145859.1 |
NP_665858.1
|
4, 5 |
405 |
|
NM_145860.1 |
NP_665859.1
|
3, 4 |
311 |
|
XM_005247086.4 |
XP_005247143.1
|
4, 5 |
1291 |
|
XM_005247087.4 |
XP_005247144.1
|
4, 5 |
1296 |
|
XM_005247088.3 |
XP_005247145.1
|
3, 4 |
387 |
|
XM_006713485.3 |
XP_006713548.1
|
4, 5 |
451 |
|
XM_011512368.2 |
XP_011510670.1
|
5, 6 |
1364 |
|
XM_011512369.2 |
XP_011510671.1
|
5, 6 |
1369 |
|
XM_017005644.1 |
XP_016861133.1
|
4, 5 |
446 |
|
GenBank mRNA |
AF022385.1 |
- |
3, 4 |
311 |
|
AK291130.1 |
- |
3, 4 |
342 |
|
BC002506.1 |
- |
4, 5 |
568 |
3050540 |
BC016353.1 |
- |
4, 5 |
409 |
4092244 |
CR457107.1 |
- |
2, 3 |
158 |
|
View Full Product Details