Gene Transcripts

Gene Symbol
Gbgt1
Entrez Gene ID
65270
Gene Name
globoside alpha-1,3-N-acetylgalactosaminyltransferase 1
Gene Aliases
Abo, Abol1
Chromosome Location
Chr. 3: 5109880 - 5126901 on Build Rnor_6.0
UniGene ID
Rn.229737
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB081649.1 - 4 235
AB081650.1 - 4 235
AB081651.1 - 3 210
AF296762.1 - 266
AF469946.1 - 3 160

View Full Product Details