Gene Transcripts
- Gene Symbol
- Sh2b3
- Entrez Gene ID
- 58838
- Gene Name
- SH2B adaptor protein 3
- Gene Aliases
- Lnk
- Chromosome Location
- Chr. 12: 40261990 - 40265757 on Build Rnor_6.0
- UniGene ID
- Rn.11228
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031621.1 |
NP_113809.1
|
2 |
931 |
|
GenBank mRNA |
U24655.1 |
- |
2 |
931 |
|
View Full Product Details