Gene Transcripts

Gene Symbol
Coq7
Entrez Gene ID
25249
Gene Name
coenzyme Q7 homolog, ubiquinone (yeast)
Gene Aliases
Chromosome Location
Chr. 1: 188176060 - 188190874 on Build Rnor_6.0
UniGene ID
Rn.6981
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012785.3 NP_036917.2 3 362
XM_006230108.2 XP_006230170.1 3 436
GenBank mRNA FQ225865.1 - 362
U46149.1 - 2 188

View Full Product Details