Gene Transcripts

Gene Symbol
PVR
Entrez Gene ID
25066
Gene Name
poliovirus receptor
Gene Aliases
Taa1, Tage4
Chromosome Location
Chr. 1: 80820306 - 80835712 on Build Rnor_6.0
UniGene ID
Rn.10677
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017076.2 NP_058772.2 2 408
GenBank mRNA BC085741.1 - 2 408
L12025.1 - 2 397

View Full Product Details