Gene Transcripts
- Gene Symbol
- TRAM1
- Entrez Gene ID
- 23471
- Gene Name
- translocation associated membrane protein 1
- Gene Aliases
- PNAS8, TRAM, TRAMP
- Chromosome Location
- Chr. 8: 70573218 - 70608459 on Build GRCh38
- UniGene ID
- Hs.491988
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001317804.1 |
NP_001304733.1
|
9, 10 |
930 |
|
NM_001317805.1 |
NP_001304734.1
|
8, 9 |
1445 |
|
NM_014294.5 |
NP_055109.1
|
8, 9 |
1013 |
|
XM_017013266.1 |
XP_016868755.1
|
7, 8 |
885 |
|
XM_017013267.1 |
XP_016868756.1
|
7, 8 |
1229 |
|
GenBank mRNA |
AK289397.1 |
- |
8, 9 |
873 |
|
AK292777.1 |
- |
8, 9 |
797 |
|
AK303411.1 |
- |
9, 10 |
858 |
|
BC000687.2 |
- |
8, 9 |
835 |
3347823 |
BC020283.1 |
- |
8, 9 |
828 |
|
BC037738.1 |
- |
8, 9 |
925 |
5261490 |
BT007359.1 |
- |
8, 9 |
753 |
|
CR541654.1 |
- |
8, 9 |
753 |
|
CR541739.1 |
- |
8, 9 |
753 |
|
X63679.1 |
- |
8, 9 |
874 |
|
View Full Product Details