Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014391.2 | NP_055206.2 | 2 | 382 | |
GenBank mRNA | AK290960.1 | - | 2 | 194 | |
AU139209.1 | - | 2 | 384 | ||
AY903446.1 | - | 2 | 151 | ||
BC005191.1 | - | 2 | 195 | ||
BC018667.1 | - | 2 | 198 | 4245591 | |
X83703.1 | - | 2 | 383 |