Gene Transcripts

Gene Symbol
SLC16A12
Entrez Gene ID
387700
Gene Name
solute carrier family 16 member 12
Gene Aliases
CJMG, CRT2, MCT12
Chromosome Location
Chr. 10: 89430294 - 89536028 on Build GRCh38
UniGene ID
Hs.530338
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_213606.3 NP_998771.3 4 555
XM_017016237.1 XP_016871726.1 3 645
XM_017016238.1 XP_016871727.1 3 584
XM_017016239.1 XP_016871728.1 3 443
GenBank mRNA AK127303.1 - 4 556
CR936657.1 - 3 414

View Full Product Details