Gene Transcripts

Gene Symbol
GSX1
Entrez Gene ID
219409
Gene Name
GS homeobox 1
Gene Aliases
GSH1, Gsh-1
Chromosome Location
Chr. 13: 27792643 - 27793952 on Build GRCh38
UniGene ID
Hs.351785
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145657.2 NP_663632.1 2 601
GenBank mRNA AB044157.1 - 2 601

View Full Product Details