Gene Transcripts

Gene Symbol
Thap1
Entrez Gene ID
306547
Gene Name
THAP domain containing, apoptosis associated protein 1
Gene Aliases
Chromosome Location
Chr. 16: 70661360 - 70665831 on Build Rnor_6.0
UniGene ID
Rn.52291
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008340.1 NP_001008341.1 3 697
GenBank mRNA BC086347.1 - 3 697
JF423321.1 - 2 501

View Full Product Details