Gene Transcripts

Gene Symbol
Rtcd1
Entrez Gene ID
295395
Gene Name
RNA terminal phosphate cyclase domain 1
Gene Aliases
Chromosome Location
Chr. 2: 219537213 - 219558489 on Build Rnor_6.0
UniGene ID
Rn.25445
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004227.1 NP_001004227.1 6 581
XM_008761456.1 XP_008759678.1 5 537
GenBank mRNA BC079374.1 - 6 581

View Full Product Details