Gene Transcripts

Gene Symbol
Emcn
Entrez Gene ID
295490
Gene Name
endomucin
Gene Aliases
Chromosome Location
Chr. 2: 242634314 - 242714190 on Build Rnor_6.0
UniGene ID
Rn.8542
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004228.1 NP_001004228.1 12 995
XM_006233366.2 XP_006233428.1 13 1101
GenBank mRNA BC079155.1 - 12 995

View Full Product Details