Gene Transcripts

Gene Symbol
Mettl10
Entrez Gene ID
72096
Gene Name
methyltransferase like 10
Gene Aliases
2010208K18Rik
Chromosome Location
Chr. 7: 132827232 - 132852880 on Build GRCm38
UniGene ID
Mm.28631
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028095.1 NP_082371.1 4 423
GenBank mRNA AK008476.1 - 4 408
BC038323.1 - 4 378
BC096622.1 - 4 407 30445095
BC116375.2 - 4 421 40101137

View Full Product Details