Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001204514.1 | NP_001191443.1 | 3 | 498 | |
NM_001204516.1 | NP_001191445.1 | 2 | 366 | ||
NM_001204517.1 | NP_001191446.1 | 3 | 1607 | ||
NM_014970.3 | NP_055785.2 | 3 | 498 | ||
XM_005244970.1 | XP_005245027.1 | 3 | 501 | ||
XM_011509307.2 | XP_011507609.2 | 3 | 501 | ||
GenBank mRNA | AK026471.1 | - | 3 | 277 | |
AK122897.1 | - | 3 | 1609 | ||
AK297368.1 | - | 2 | 293 | ||
AK303052.1 | - | 3 | 455 | ||
AK315586.1 | - | 3 | 412 | ||
BC028679.2 | - | 3 | 455 | 4829354 | |
BC048012.1 | - | 2 | 410 | ||
BG707718.1 | - | 3 | 507 | ||
DA144269.1 | - | 3 | 501 | ||
U59919.1 | - | 3 | 415 |