Gene Transcripts

Gene Symbol
GSTTP1
Entrez Gene ID
25774
Gene Name
glutathione S-transferase theta pseudogene 1
Gene Aliases
GSTT4, HS322B1A
Chromosome Location
Chr. 22: 23998401 - 24005361 on Build GRCh38
UniGene ID
Hs.652490
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003081.1 1, 2 125
GenBank mRNA AL050257.1 - 1, 2 125
BC096726.1 - 1, 2 125
BC098364.1 - 1, 2 125
BC099710.1 - 1, 2 125

View Full Product Details