Gene Transcripts
- Gene Symbol
- SNX9
- Entrez Gene ID
- 51429
- Gene Name
- sorting nexin 9
- Gene Aliases
- SDP1, SH3PX1, SH3PXD3A, WISP
- Chromosome Location
- Chr. 6: 157716046 - 157945077 on Build GRCh38
- UniGene ID
- Hs.191213
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016224.4 |
NP_057308.1
|
18 |
4082 |
|
GenBank mRNA |
AF172847.1 |
- |
18 |
3966 |
|
View Full Product Details