Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001127889.4 | NP_001121361.1 | 3 | 261 | |
NM_001127890.4 | NP_001121362.1 | 3 | 442 | ||
NM_016630.6 | NP_057714.1 | 3 | 442 | ||
XM_005254437.4 | XP_005254494.1 | 4 | 475 | ||
XM_017022297.1 | XP_016877786.1 | 3 | 810 | ||
XM_017022298.1 | XP_016877787.1 | 4 | 900 | ||
GenBank mRNA | AF208861.1 | - | 2 | 157 | |
AF212231.1 | - | 3 | 229 | ||
AK123725.1 | - | 1 | 674 | ||
AK172849.1 | - | 3 | 274 | ||
AK225527.1 | - | 3 | 274 | ||
AK301362.1 | - | 3 | 407 | ||
AK312943.1 | - | 3 | 276 | ||
BC000244.1 | - | 3 | 312 | 3357388 | |
BI552731.1 | - | 3 | 412 | ||
BI554234.1 | - | 4 | 470 | ||
BI771887.1 | - | 3 | 276 | ||
BP268459.1 | - | 3 | 435 | ||
BP268875.1 | - | 3 | 435 | ||
BX446863.2 | - | 3 | 392 | ||
DA680695.1 | - | 3 | 260 |