Gene Transcripts

Gene Symbol
Olr6
Entrez Gene ID
365175
Gene Name
olfactory receptor 6
Gene Aliases
Chromosome Location
Chr. 1: 70480941 - 70481879 on Build Rnor_6.0
UniGene ID
Rn.142003
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000539.1 NP_001000539.1 1 901

Gene Symbol
LOC100910456
Entrez Gene ID
100910456
Gene Name
olfactory receptor 5-like
Gene Aliases
Chromosome Location
Chr. 1: 71092733 - 71093395 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003748751.2 XP_003748799.2 1 625

View Full Product Details