Gene Transcripts

Gene Symbol
COMMD4
Entrez Gene ID
54939
Gene Name
COMM domain containing 4
Gene Aliases
Chromosome Location
Chr. 15: 75335996 - 75341927 on Build GRCh38
UniGene ID
Hs.351327
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001284377.1 NP_001271306.1 3 210
NM_001284378.1 NP_001271307.1 3 210
NM_001284379.1 NP_001271308.1 2 188
NM_001321844.1 NP_001308773.1 3 204
NM_001321848.1 NP_001308777.1 2 188
NM_017828.4 NP_060298.2 3 210
NR_104312.1 3 210
NR_104313.1 3 210
XM_005254509.3 XP_005254566.1 3 236
XM_011521739.1 XP_011520041.1 3 245
XM_011521741.1 XP_011520043.1 3 245
GenBank mRNA AK000459.1 - 3 147
AK057145.1 - 2 363
AK225716.1 - 3 171
AK293708.1 - 3 146
AK314740.1 - 3 143
AL136872.1 - 3 130
AY542160.1 - 2 116
BC000837.2 - 2 113
BC033744.1 - 2 382
BC068998.1 - 3 132 6586373
BC069195.1 - 3 138
BI829264.1 - 3 186
BI829484.1 - 3 210
BQ233598.1 - 173
BU540913.1 - 3 152
CR533452.1 - 2 116
HY162014.1 - 2 189

Gene Symbol
GOLGA6B
Entrez Gene ID
55889
Gene Name
golgin A6 family member B
Gene Aliases
GOLGA, GOLGA6D
Chromosome Location
Chr. 15: 72638291 - 72669604 on Build GRCh38
UniGene ID
Hs.546408
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017022406.1 XP_016877895.1 2 646

Gene Symbol
LOC440292
Entrez Gene ID
440292
Gene Name
COMM domain-containing protein 4-like
Gene Aliases
Chromosome Location
Chr. 15: 77938221 - 77944561 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011522301.2 XP_011520603.1 2 220
XM_017022799.1 XP_016878288.1 2 640

Gene Symbol
LOC732265
Entrez Gene ID
732265
Gene Name
uncharacterized LOC732265
Gene Aliases
Chromosome Location
Chr. 15: 74088153 - 74100467 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017022776.1 XP_016878265.1 3 558

View Full Product Details