Gene Transcripts
- Gene Symbol
- MT1DP
- Entrez Gene ID
- 326343
- Gene Name
- metallothionein 1D, pseudogene
- Gene Aliases
- MTM
- Chromosome Location
- Chr. 16: 56643687 - 56644941 on Build GRCh38
- UniGene ID
- Hs.679600
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF348999.2 |
- |
4 |
290 |
|
View Full Product Details