Gene Transcripts

Gene Symbol
Mrgprb5
Entrez Gene ID
404239
Gene Name
MAS-related GPR, member B5
Gene Aliases
Mgrg11, MrgB5
Chromosome Location
Chr. 7: 48168017 - 48168985 on Build GRCm38
UniGene ID
Mm.377927
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207538.1 NP_997421.1 1 147
GenBank mRNA BC120849.1 - 1 180
BC132003.1 - 1 181 40130280

View Full Product Details