Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001014057.1 | NP_001014079.1 | 9 | 1154 | |
XM_008763240.1 | XP_008761462.1 | 9 | 1177 | ||
XM_008763241.1 | XP_008761463.1 | 9 | 1216 | ||
XM_008763242.1 | XP_008761464.1 | 9 | 1178 | ||
GenBank mRNA | BC087631.1 | - | 9 | 1154 |