Gene Transcripts

Gene Symbol
SPINK13
Entrez Gene ID
153218
Gene Name
serine peptidase inhibitor, Kazal type 13 (putative)
Gene Aliases
HBVDNAPTP1, HESPINTOR, LiESP6, SPINK5L3
Chromosome Location
Chr. 5: 148268860 - 148286210 on Build GRCh38
UniGene ID
Hs.483771
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040129.2 NP_001035218.1 4, 5 306
GenBank mRNA AK001520.1 - 5, 6 775
BC128163.1 - 5, 6 458 40113027
BC128164.1 - 4, 5 306 40113028
DQ438947.1 - 3, 4 244
EF108324.1 - 4, 5 354
EF426754.1 - 4, 5 354

View Full Product Details