Gene Transcripts

Gene Symbol
Defb27
Entrez Gene ID
641642
Gene Name
defensin beta 27
Gene Aliases
Chromosome Location
Chr. 3: 148069922 - 148073554 on Build Rnor_6.0
UniGene ID
Rn.117334
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037519.1 NP_001032608.1 2 306
GenBank mRNA AY621358.1 - 2 306

View Full Product Details