Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025227.2 | NP_079503.1 | 3 | 375 | |
GenBank mRNA | AF465765.1 | - | 2 | 158 | |
AK027068.1 | - | 3 | 352 | ||
AK172819.1 | - | 3 | 354 | ||
AK304064.1 | - | 3 | 280 | ||
AY358723.1 | - | 3 | 283 | ||
BC034415.1 | - | 3 | 229 | 5173032 | |
BP333980.1 | - | 3 | 374 |