Gene Transcripts

Gene Symbol
PDRG1
Entrez Gene ID
81572
Gene Name
p53 and DNA damage regulated 1
Gene Aliases
C20orf126, PDRG
Chromosome Location
Chr. 20: 31944955 - 31952080 on Build GRCh38
UniGene ID
Hs.435755
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030815.2 NP_110442.1 2 237
GenBank mRNA AK312021.1 - 2 181
AY286301.1 - 2 249
BC001856.2 - 2 108
BC009334.2 - 2 162 4126826

View Full Product Details