Gene Transcripts
- Gene Symbol
- SLC2A11
- Entrez Gene ID
- 66035
- Gene Name
- solute carrier family 2 member 11
- Gene Aliases
- GLUT10, GLUT11
- Chromosome Location
- Chr. 22: 23856703 - 23886112 on Build GRCh38
- UniGene ID
- Hs.661910
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001024938.3 |
NP_001020109.1
|
3, 4 |
557 |
|
NM_001024939.3 |
NP_001020110.1
|
3, 4 |
445 |
|
NM_001282864.1 |
NP_001269793.1
|
3, 4 |
800 |
|
NM_030807.4 |
NP_110434.3
|
4, 5 |
668 |
|
NR_104248.1 |
|
4, 5 |
668 |
|
GenBank mRNA |
AB209301.1 |
- |
2, 3 |
203 |
|
AF443201.1 |
- |
4, 5 |
499 |
|
AF479409.1 |
- |
2, 3 |
238 |
|
AJ271290.1 |
- |
3, 4 |
388 |
|
AK075467.1 |
- |
3, 4 |
333 |
|
AK300196.1 |
- |
3, 4 |
800 |
|
AK304409.1 |
- |
3, 4 |
423 |
|
AK314502.1 |
- |
4, 5 |
439 |
|
BC063525.1 |
- |
4, 5 |
425 |
|
BC094735.1 |
- |
3, 4 |
374 |
5304025 |
BC100808.2 |
- |
3, 4 |
303 |
40000757 |
BC100809.1 |
- |
3, 4 |
303 |
40000760 |
CR456373.1 |
- |
3, 4 |
334 |
|
DC303452.1 |
- |
3, 4 |
446 |
|
View Full Product Details