Gene Transcripts
- Gene Symbol
- KIF6
- Entrez Gene ID
- 221458
- Gene Name
- kinesin family member 6
- Gene Aliases
- C6orf102, dJ1043E3.1, dJ137F1.4, dJ188D3.1
- Chromosome Location
- Chr. 6: 39335100 - 39725405 on Build GRCh38
- UniGene ID
- Hs.588202
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001289020.1 |
NP_001275949.1
|
16, 17 |
1984 |
|
NM_001289021.1 |
NP_001275950.1
|
16, 17 |
1867 |
|
NM_001289024.1 |
NP_001275953.1
|
4, 5 |
508 |
|
NM_145027.4 |
NP_659464.3
|
17, 18 |
2035 |
|
XM_005248904.4 |
XP_005248961.1
|
17, 18 |
2035 |
|
XM_011514357.2 |
XP_011512659.1
|
17, 18 |
1960 |
|
XM_011514358.2 |
XP_011512660.1
|
17, 18 |
1960 |
|
XM_011514359.2 |
XP_011512661.1
|
17, 18 |
1960 |
|
XM_017010427.1 |
XP_016865916.1
|
16, 17 |
1925 |
|
XM_017010428.1 |
XP_016865917.1
|
12, 13 |
1385 |
|
GenBank mRNA |
AK123217.1 |
- |
4, 5 |
508 |
|
AK131471.1 |
- |
17, 18 |
2035 |
|
AL832634.2 |
- |
4, 5 |
271 |
|
BC022074.2 |
- |
4, 5 |
449 |
|
BC098418.1 |
- |
4, 5 |
467 |
|
BC117307.1 |
- |
16, 17 |
1797 |
40125858 |
BC143444.1 |
- |
16, 17 |
1914 |
9051952 |
BC143445.1 |
- |
16, 17 |
1797 |
9051953 |
BX649045.1 |
- |
14, 15 |
1615 |
|
DA167995.1 |
- |
4, 5 |
508 |
|
View Full Product Details