Gene Transcripts

Gene Symbol
TMEM199
Entrez Gene ID
147007
Gene Name
transmembrane protein 199
Gene Aliases
C17orf32, CDG2P, VMA12, VPH2
Chromosome Location
Chr. 17: 28357581 - 28363683 on Build GRCh38
UniGene ID
Hs.594696
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152464.2 NP_689677.1 6 1498
GenBank mRNA AK311363.1 - 1838
BC033113.1 - 6 1415 5429060
BQ891740.1 - 1 243

View Full Product Details