Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001030.4 | NP_001021.1 | 1 | 237 | |
GenBank mRNA | AK312070.1 | - | 3 | 238 | |
AV763942.1 | - | 1 | 238 | ||
BC002658.2 | - | 1 | 220 | 3607957 | |
BC070219.1 | - | 238 | 6744779 | ||
CN304370.1 | - | 1 | 233 | ||
L19739.1 | - | 3 | 223 | ||
U57847.1 | - | 3 | 216 |