Gene Transcripts

Gene Symbol
Tmprss11a
Entrez Gene ID
194597
Gene Name
transmembrane protease, serine 11a
Gene Aliases
Gm7
Chromosome Location
Chr. 5: 86410410 - 86469175 on Build GRCm38
UniGene ID
Mm.249106
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033233.2 NP_001028405.1 4 377
GenBank mRNA AK142820.1 - 4 378

View Full Product Details