Gene Transcripts

Gene Symbol
Vmn2r80
Entrez Gene ID
624765
Gene Name
vomeronasal 2, receptor 80
Gene Aliases
EG624765
Chromosome Location
Chr. 10: 79148816 - 79194933 on Build GRCm38
UniGene ID
Mm.469843
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001103368.1 NP_001096838.1 6 2140

View Full Product Details