Gene Transcripts

Gene Symbol
Car4
Entrez Gene ID
29242
Gene Name
carbonic anhydrase 4
Gene Aliases
Ca4
Chromosome Location
Chr. 10: 72272286 - 72281069 on Build Rnor_6.0
UniGene ID
Rn.51389
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019174.3 NP_062047.1 7 826
GenBank mRNA BC097329.1 - 7 721
CB746957.1 - 4 387
S68245.1 - 7 769

View Full Product Details