Gene Transcripts

Gene Symbol
Fra10ac1
Entrez Gene ID
70567
Gene Name
FRA10AC1 homolog (human)
Gene Aliases
5730455O13Rik, AI448293
Chromosome Location
Chr. 19: 38188477 - 38224513 on Build GRCm38
UniGene ID
Mm.30607
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081075.1 NP_001074544.1 14 1318
GenBank mRNA AK077572.1 - 14 1291
AK172241.1 - 14 1325

View Full Product Details