Gene Transcripts
- Gene Symbol
- Axdnd1
- Entrez Gene ID
- 77352
- Gene Name
- axonemal dynein light chain domain containing 1
- Gene Aliases
- 9430070O13Rik, Gm979
- Chromosome Location
- Chr. 1: 156330415 - 156420915 on Build GRCm38
- UniGene ID
- Mm.428709
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK134071.1 |
- |
11 |
1605 |
|
View Full Product Details