Gene Transcripts

Gene Symbol
Hcrtr1
Entrez Gene ID
25593
Gene Name
hypocretin (orexin) receptor 1
Gene Aliases
Hctr1
Chromosome Location
Chr. 5: 148346029 - 148355404 on Build Rnor_6.0
UniGene ID
Rn.88262
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013064.1 NP_037196.1 3 702
GenBank mRNA AF041244.1 - 3 702

View Full Product Details