Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009225.2 | NP_033251.1 | 3 | 318 | |
GenBank mRNA | AK077537.1 | - | 3 | 271 | |
AK146319.1 | - | 3 | 361 | ||
AK178243.1 | - | 2 | 118 | ||
AK186005.1 | - | 2 | 118 | ||
AK203845.1 | - | 2 | 118 | ||
AK207139.1 | - | 2 | 118 | ||
BG086527.2 | - | 3 | 276 | ||
BY280308.1 | - | 3 | 318 |