Gene Transcripts

Gene Symbol
Gsto1
Entrez Gene ID
14873
Gene Name
glutathione S-transferase omega 1
Gene Aliases
AA407097, AI194287, AU018802, GSTO 1-1, GSTX, SPG-R, p28
Chromosome Location
Chr. 19: 47854989 - 47864790 on Build GRCm38
UniGene ID
Mm.378931
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010362.2 NP_034492.1 4 516
GenBank mRNA AK027922.1 - 4 466
AK146834.1 - 4 478
AK168383.1 - 4 478
BC085165.1 - 4 454 30359900
U80819.1 - 4 516

View Full Product Details