Gene Transcripts

Gene Symbol
Jag2
Entrez Gene ID
16450
Gene Name
jagged 2
Gene Aliases
D12Ggc2e, Serh, mJagged2-1, sm
Chromosome Location
Chr. 12: 112907803 - 112929764 on Build GRCm38
UniGene ID
Mm.186146
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010588.2 NP_034718.2 10 1304
GenBank mRNA AF010137.1 - 6 533
AF038572.1 - 10 1304
Y14495.1 - 4 322

View Full Product Details