Gene Transcripts

Gene Symbol
Hnrnpa2b1
Entrez Gene ID
53379
Gene Name
heterogeneous nuclear ribonucleoprotein A2/B1
Gene Aliases
9130414A06Rik, Hnrpa2, Hnrpa2b1, hnrnp-A
Chromosome Location
Chr. 6: 51460434 - 51470492 on Build GRCm38
UniGene ID
Mm.155896
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016806.3 NP_058086.2 3 377
NM_182650.4 NP_872591.1 3 377
GenBank mRNA AF073993.1 - 3 255
AF406651.2 - 2 166
AK049771.1 - 2 2742
AK089012.1 - 3 328
AK137214.1 - 3 378
BB627924.1 - 3 378
BC141253.1 - 3 211 9055865
BC145588.1 - 3 211 9054154

View Full Product Details