Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039137.3 | NP_001034226.1 | 4 | 1210 | |
NM_001285992.1 | NP_001272921.1 | 4 | 1312 | ||
NM_019708.4 | NP_062682.1 | 4 | 1315 | ||
GenBank mRNA | AF115778.1 | - | 4 | 1137 | |
AK002521.1 | - | 4 | 1160 | ||
AK076085.1 | - | 4 | 1108 | ||
AK167488.1 | - | 4 | 1158 | ||
BC017629.1 | - | 4 | 1134 | ||
BC085265.1 | - | 4 | 1121 | 6391098 | |
BC092071.1 | - | 4 | 1118 | 4505111 | |
BC108407.1 | - | 4 | 1138 |