Gene Transcripts
- Gene Symbol
- Sh3kbp1
- Entrez Gene ID
- 58194
- Gene Name
- SH3-domain kinase binding protein 1
- Gene Aliases
- 1200007H22Rik, 1700125L08Rik, 5830464D22Rik, AI447724, IN85, Ruk, Seta
- Chromosome Location
- Chr. X: 159627029 - 159975920 on Build GRCm38
- UniGene ID
- Mm.286495
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135727.2 |
NP_001129199.1
|
4, 5 |
802 |
|
NM_021389.6 |
NP_067364.2
|
3, 4 |
367 |
|
GenBank mRNA |
AK004636.1 |
- |
3, 4 |
367 |
|
AK018032.1 |
- |
3, 4 |
383 |
|
AK020782.1 |
- |
3, 4 |
344 |
|
AK030839.1 |
- |
3, 4 |
368 |
|
AK038540.1 |
- |
4, 5 |
534 |
|
AK049182.1 |
- |
4, 5 |
803 |
|
AK080930.1 |
- |
4, 5 |
667 |
|
AK172022.1 |
- |
4, 5 |
783 |
|
BC034646.1 |
- |
3, 4 |
371 |
|
BC052863.1 |
- |
3, 4 |
316 |
|
View Full Product Details