Gene Transcripts
- Gene Symbol
- Rtn4
- Entrez Gene ID
- 68585
- Gene Name
- reticulon 4
- Gene Aliases
- 1110020G17Rik, AA407876, AA409940, AA960376, ASY, C130026I10Rik, NOGO, NSP-CL, NgA, Nogo-A, Nogo-B, Nogo-C, mKIAA0886, mKIAA4153
- Chromosome Location
- Chr. 11: 29692898 - 29744414 on Build GRCm38
- UniGene ID
- Mm.192580
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024226.4 |
NP_077188.1
|
2, 3 |
536 |
|
NM_194051.3 |
NP_918940.1
|
4, 5 |
2823 |
|
NM_194052.3 |
NP_918941.1
|
2, 3 |
1012 |
|
NM_194053.3 |
NP_918942.1
|
3, 4 |
1069 |
|
NM_194054.3 |
NP_918943.1
|
4, 5 |
3430 |
|
GenBank mRNA |
AB073672.1 |
- |
2, 3 |
1388 |
|
AF326337.1 |
- |
2, 3 |
316 |
|
AK034902.1 |
- |
2, 3 |
956 |
|
AK211699.1 |
- |
2, 3 |
252 |
|
AK212390.1 |
- |
2, 3 |
239 |
|
AK220511.1 |
- |
4, 5 |
3390 |
|
AY102280.1 |
- |
4, 5 |
2823 |
|
AY102281.1 |
- |
2, 3 |
969 |
|
AY102282.1 |
- |
3, 4 |
1026 |
|
AY102283.1 |
- |
2, 3 |
529 |
|
AY102284.1 |
- |
4, 5 |
3387 |
|
AY114152.1 |
- |
4, 5 |
3388 |
|
AY114153.1 |
- |
3, 4 |
843 |
|
BC032192.1 |
- |
2, 3 |
1682 |
5323152 |
BC032272.1 |
- |
2, 3 |
2536 |
|
BC056373.1 |
- |
4, 5 |
3258 |
|
BU848611.1 |
- |
2, 3 |
375 |
|
View Full Product Details