Gene Transcripts

Gene Symbol
Snrnp27
Entrez Gene ID
66618
Gene Name
small nuclear ribonucleoprotein 27 (U4/U6.U5)
Gene Aliases
2610209M04Rik, AI449063, AU022368, Ry1
Chromosome Location
Chr. 6: 86675169 - 86684491 on Build GRCm38
UniGene ID
Mm.182650
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025665.2 NP_079941.1 4 301
GenBank mRNA AK011937.1 - 4 302
AK045231.1 - 4 314
AK160190.1 - 4 311
BC027564.1 - 4 302 3468252

View Full Product Details