Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000263.3 | NP_000254.2 | 3 | 906 | |
XM_006721920.2 | XP_006721983.1 | 3 | 299 | ||
XM_011524840.1 | XP_011523142.1 | 3 | 299 | ||
XM_017024686.1 | XP_016880175.1 | 2 | 439 | ||
XM_017024687.1 | XP_016880176.1 | 2 | 643 | ||
GenBank mRNA | BC053991.1 | - | 3 | 589 | |
L78464.1 | - | 3 | 572 | ||
U40846.1 | - | 3 | 667 | ||
U43573.1 | - | 3 | 566 |