Gene Transcripts

Gene Symbol
Slc46a3
Entrez Gene ID
71706
Gene Name
solute carrier family 46, member 3
Gene Aliases
1200006F02Rik
Chromosome Location
Chr. 5: 147877846 - 147894864 on Build GRCm38
UniGene ID
Mm.153218
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027872.3 NP_082148.1 2 456
GenBank mRNA AK004609.1 - 2 241
AK156898.1 - 2 458
BC006902.1 - 2 237 3599960

View Full Product Details